The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166271.2 | NP_001159743.1 | 4 - 5 | 2489 | 59 |
NM_001286792.1 | NP_001273721.1 | 6 - 7 | 2813 | 59 | |
NM_153023.3 | NP_694568.1 | 3 - 4 | 725 | 59 | |
GenBank mRNA | AK055770.1 | - | 3 - 4 | 693 | 59 |
AK299981.1 | - | 1 - 2 | 166 | 59 | |
AK301149.1 | - | 1 - 2 | 166 | 59 | |
BC109290.1 | - | 2 - 3 | 435 | 59 | |
BC109291.1 | - | 2 - 3 | 435 | 59 | |
BX648244.1 | - | 6 - 7 | 2635 | 59 |