Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCGB2B2
Entrez Gene ID
284402
Gene Name
secretoglobin family 2B member 2
Gene Aliases
SCGB4A2, SCGBL
Chromosome Location
Chr.19: 34590625 - 34594585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025591.3 NP_001020762.1 1 - 2 81 73
GenBank mRNA AK093495.1 - 2 - 3 2146 73
BC093909.1 - 1 - 2 81 73
BC093935.1 - 1 - 2 81 73

View Full Product Details