The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207348.2 | NP_997231.1 | 2 - 3 | 615 | 102 |
XM_011541292.2 | XP_011539594.1 | 2 - 3 | 1841 | 102 | |
XM_011541293.1 | XP_011539595.1 | 2 - 3 | 1841 | 102 | |
XM_017001083.1 | XP_016856572.1 | 2 - 3 | 1841 | 102 | |
GenBank mRNA | BC027998.1 | - | 2 - 3 | 615 | 102 |