Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A34
Entrez Gene ID
284723
Gene Name
solute carrier family 25 member 34
Gene Aliases
-
Chromosome Location
Chr.1: 15735088 - 15741392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207348.2 NP_997231.1 3 - 4 769 96
XM_011541292.2 XP_011539594.1 3 - 4 1995 96
XM_011541293.1 XP_011539595.1 3 - 4 1995 96
GenBank mRNA BC027998.1 - 3 - 4 769 96

View Full Product Details