Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC69
Entrez Gene ID
100130742
Gene Name
leucine rich repeat containing 69
Gene Aliases
-
Chromosome Location
Chr.8: 91102619 - 91219236 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK097393.1 - 11 - 12 1491 69

View Full Product Details