Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR97
Entrez Gene ID
340390
Gene Name
WD repeat domain 97
Gene Aliases
KIAA1875
Chromosome Location
Chr.8: 144107692 - 144116392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316309.1 NP_001303238.1 8 - 9 2229 86
XM_017013385.1 XP_016868874.1 8 - 9 2223 86
XM_017013386.1 XP_016868875.1 8 - 9 2223 86
XM_017013387.1 XP_016868876.1 8 - 9 2164 86
XM_017013388.1 XP_016868877.1 8 - 9 2223 86
XM_017013389.1 XP_016868878.1 8 - 9 2223 86
XM_017013390.1 XP_016868879.1 8 - 9 2223 86
XM_017013391.1 XP_016868880.1 8 - 9 2223 86
XM_017013392.1 XP_016868881.1 8 - 9 2223 86
XM_017013393.1 XP_016868882.1 8 - 9 2223 86
XM_017013394.1 XP_016868883.1 7 - 8 2009 86
GenBank mRNA AB058778.1 - 8 - 9 2195 86
AK095597.1 - 2 - 3 961 86


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream MAF1 homolog, negative regulator of RNA polymerase III MAF1 -3.0kb Sense Chr. - 144104279 - 144107612 on Build GRCh38
Downstream HGH1 homolog HGH1 BRP16, BRP16L, C8orf30A, C8orf30B, FAM203A, FAM203B 26.8kb Sense Chr. - 144137721 - 144140843 on Build GRCh38

View Full Product Details