The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138382.2 | NP_612391.1 | 1 - 2 | 200 | 115 |
GenBank mRNA | BC007652.1 | - | 1 - 2 | 186 | 115 |
BC101998.1 | - | 1 - 2 | 200 | 115 | |
BC105691.1 | - | 1 - 2 | 200 | 115 | |
BC105692.1 | - | 1 - 2 | 200 | 115 |