The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138382.2 | NP_612391.1 | 3 - 4 | 337 | 63 |
GenBank mRNA | BC007652.1 | - | 3 - 4 | 323 | 63 |
BC101998.1 | - | 3 - 4 | 337 | 63 | |
BC105691.1 | - | 3 - 4 | 337 | 63 | |
BC105692.1 | - | 3 - 4 | 337 | 63 | |
BC110436.1 | - | 2 - 3 | 1254 | 63 |