The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014008.4 | NP_054727.1 | 7 - 8 | 1079 | 70 |
XM_005272599.3 | XP_005272656.1 | 7 - 8 | 1008 | 70 | |
GenBank mRNA | AJ005890.1 | - | 7 - 8 | 995 | 70 |
AK291976.1 | - | 7 - 8 | 1040 | 70 | |
AK298264.1 | - | 7 - 8 | 1074 | 70 | |
AK316375.1 | - | 7 - 8 | 1025 | 70 | |
BC000972.2 | - | 7 - 8 | 1026 | 70 | |
BC011675.2 | - | 7 - 8 | 910 | 70 |