The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199570.1 | NP_001186499.1 | 12 - 13 | 1902 | 75 |
NM_015526.2 | NP_056341.1 | 13 - 14 | 1818 | 75 | |
GenBank mRNA | AJ427922.1 | - | 13 - 14 | 1796 | 75 |
AK056445.1 | - | 11 - 12 | 1301 | 75 | |
AK090732.1 | - | 13 - 14 | 1821 | 75 | |
AK092354.1 | - | 9 - 10 | 1065 | 75 | |
AK094738.1 | - | 13 - 14 | 1816 | 75 | |
AK289509.1 | - | 12 - 13 | 1870 | 75 | |
AK294748.1 | - | 13 - 14 | 1689 | 75 | |
AL117468.1 | - | 5 - 6 | 430 | 75 | |
BC013116.1 | - | 13 - 14 | 1730 | 75 | |
BC014486.1 | - | 10 - 11 | 1261 | 75 |