The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145648.1 | NP_001139120.1 | 12 - 13 | 2116 | 75 |
NM_002891.4 | NP_002882.3 | 12 - 13 | 2116 | 75 | |
XM_011521866.2 | XP_011520168.1 | 12 - 13 | 2016 | 75 | |
XM_011521867.2 | XP_011520169.1 | 12 - 13 | 2016 | 75 | |
XM_017022455.1 | XP_016877944.1 | 12 - 13 | 2016 | 75 | |
XM_017022456.1 | XP_016877945.1 | 12 - 13 | 2016 | 75 | |
XM_017022457.1 | XP_016877946.1 | 12 - 13 | 2016 | 75 | |
XM_017022458.1 | XP_016877947.1 | 12 - 13 | 2016 | 75 | |
XM_017022459.1 | XP_016877948.1 | 12 - 13 | 2016 | 75 | |
GenBank mRNA | AK226101.1 | - | 11 - 12 | 1400 | 75 |
AK290135.1 | - | 12 - 13 | 1982 | 75 | |
AK301205.1 | - | 1 - 2 | 204 | 75 | |
AK309047.1 | - | 11 - 12 | 1538 | 75 | |
BC040275.1 | - | 12 - 13 | 2007 | 75 | |
L26584.1 | - | 12 - 13 | 1741 | 75 |