The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145648.1 | NP_001139120.1 | 15 - 16 | 2820 | 149 |
NM_002891.4 | NP_002882.3 | 16 - 17 | 2868 | 149 | |
NM_153815.2 | NP_722522.1 | 2 - 3 | 1314 | 149 | |
XM_011521866.2 | XP_011520168.1 | 16 - 17 | 2768 | 149 | |
XM_011521867.2 | XP_011520169.1 | 16 - 17 | 2768 | 149 | |
XM_017022455.1 | XP_016877944.1 | 16 - 17 | 2759 | 149 | |
XM_017022456.1 | XP_016877945.1 | 15 - 16 | 2729 | 149 | |
XM_017022457.1 | XP_016877946.1 | 15 - 16 | 2729 | 149 | |
XM_017022458.1 | XP_016877947.1 | 15 - 16 | 2720 | 149 | |
XM_017022459.1 | XP_016877948.1 | 15 - 16 | 2729 | 149 | |
GenBank mRNA | AK226101.1 | - | 14 - 15 | 2104 | 149 |
AK290135.1 | - | 16 - 17 | 2734 | 149 | |
AK301205.1 | - | 4 - 5 | 917 | 149 | |
AK309047.1 | - | 14 - 15 | 2251 | 149 | |
BC040275.1 | - | 15 - 16 | 2711 | 149 | |
L26584.1 | - | 16 - 17 | 2499 | 149 | |
S62035.1 | - | 2 - 3 | 1323 | 149 |