The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023083.3 | NP_075571.1 | 1 - 2 | 339 | 77 |
NM_023085.3 | NP_075573.2 | 1 - 2 | 339 | 77 | |
GenBank mRNA | AB058748.1 | - | 1 - 2 | 243 | 77 |
AF089088.1 | - | 1 - 2 | 320 | 77 | |
AF089090.1 | - | 1 - 2 | 184 | 77 | |
AF089091.1 | - | 1 - 2 | 184 | 77 | |
AF089092.1 | - | 1 - 2 | 184 | 77 | |
AF089093.1 | - | 1 - 2 | 184 | 77 | |
AF089094.1 | - | 1 - 2 | 184 | 77 | |
AK074807.1 | - | 1 - 2 | 290 | 77 | |
AK300279.1 | - | 1 - 2 | 321 | 77 | |
AL558905.3 | - | 1 - 2 | 337 | 77 | |
BC004260.1 | - | 1 - 2 | 288 | 77 | |
BC007553.1 | - | 1 - 2 | 288 | 77 | |
DA413480.1 | - | 1 - 2 | 330 | 77 | |
DA623373.1 | - | 1 - 2 | 327 | 77 |