The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015512.4 | NP_056327.4 | 64 - 65 | 10539 | 78 |
XM_017006129.1 | XP_016861618.1 | 66 - 67 | 13496 | 78 | |
XM_017006130.1 | XP_016861619.1 | 65 - 66 | 13426 | 78 | |
XM_017006131.1 | XP_016861620.1 | 65 - 66 | 13370 | 78 | |
GenBank mRNA | AB037831.3 | - | 64 - 65 | 10539 | 78 |
AB290163.1 | - | 63 - 64 | 10734 | 78 | |
AK023766.1 | - | 3 - 4 | 549 | 78 | |
AK074130.2 | - | 11 - 12 | 2064 | 78 | |
AY221994.1 | - | 20 - 21 | 3582 | 78 |