Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
Chromosome Location
Chr.3: 15450133 - 15521751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005677.3 NP_005668.2 6 - 7 586 115
NM_080538.2 NP_536799.1 6 - 7 715 115
NM_080539.3 NP_536800.2 5 - 6 484 115
GenBank mRNA AF057036.1 - 6 - 7 586 115
AJ225895.1 - 6 - 7 579 115
AK128401.1 - 6 - 7 715 115
AK289688.1 - 6 - 7 605 115
AY150334.1 - 6 - 7 563 115
AY150335.1 - 6 - 7 563 115
AY150336.1 - 6 - 7 563 115
AY150337.1 - 6 - 7 563 115
AY150338.1 - 6 - 7 563 115
AY150339.1 - 6 - 7 563 115
BC074828.2 - 6 - 7 490 115
BC074829.2 - 6 - 7 490 115

View Full Product Details