The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257118.2 | NP_001244047.1 | 2 - 3 | 324 | 140 |
NM_033292.3 | NP_150634.1 | 2 - 3 | 324 | 140 | |
XM_017018393.1 | XP_016873882.1 | 2 - 3 | 672 | 140 | |
XM_017018394.1 | XP_016873883.1 | 2 - 3 | 672 | 140 | |
XM_017018396.1 | XP_016873885.1 | 2 - 3 | 581 | 140 | |
GenBank mRNA | AK223503.1 | - | 2 - 3 | 316 | 140 |
AK290114.1 | - | 2 - 3 | 315 | 140 | |
AK296646.1 | - | 2 - 3 | 306 | 140 | |
AK301037.1 | - | 2 - 3 | 224 | 140 | |
AK313516.1 | - | 2 - 3 | 310 | 140 | |
AY660536.1 | - | 2 - 3 | 218 | 140 | |
M87507.1 | - | 2 - 3 | 297 | 140 | |
X65019.1 | - | 1 - 2 | 287 | 140 |