The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032805.2 | NP_116194.2 | 2 - 3 | 592 | 76 |
XM_017023791.1 | XP_016879280.1 | 1 - 2 | 628 | 76 | |
XM_017023792.1 | XP_016879281.1 | 1 - 2 | 628 | 76 | |
GenBank mRNA | AK027455.1 | - | 1 - 2 | 320 | 76 |
CX781288.1 | - | 2 - 3 | 592 | 76 |