The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033125.3 | NP_149116.2 | 2 - 3 | 595 | 136 |
XM_011536204.2 | XP_011534506.1 | 2 - 3 | 609 | 136 | |
XM_011536205.2 | XP_011534507.1 | 2 - 3 | 567 | 136 | |
XM_011536210.1 | XP_011534512.1 | 2 - 3 | 567 | 136 | |
GenBank mRNA | AB055798.1 | - | 4 - 5 | 710 | 136 |
AF268892.1 | - | 2 - 3 | 528 | 136 | |
AK057306.1 | - | 1 - 2 | 279 | 136 | |
AY145502.1 | - | 2 - 3 | 528 | 136 | |
BC037247.1 | - | 2 - 3 | 543 | 136 | |
BC047565.1 | - | 2 - 3 | 595 | 136 | |
GQ472228.1 | - | 2 - 3 | 595 | 136 |