The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014740.3 | NP_055555.1 | 9 - 10 | 1203 | 70 |
XM_011525522.1 | XP_011523824.1 | 8 - 9 | 1096 | 70 | |
GenBank mRNA | AK290608.1 | - | 9 - 10 | 1184 | 70 |
BC003662.2 | - | 9 - 10 | 1113 | 70 | |
BC004386.1 | - | 9 - 10 | 1134 | 70 | |
BC011151.1 | - | 9 - 10 | 1138 | 70 | |
CA428254.1 | - | 3 - 4 | 70 | ||
CR456750.1 | - | 9 - 10 | 981 | 70 | |
D21853.1 | - | 9 - 10 | 1195 | 70 | |
X79538.1 | - | 9 - 10 | 1054 | 70 |