The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004055.1 | NP_001004055.1 | 5 - 6 | 762 | 58 |
NM_012163.2 | NP_036295.1 | 5 - 6 | 927 | 58 | |
XM_017023126.1 | XP_016878615.1 | 5 - 6 | 1191 | 58 | |
XM_017023127.1 | XP_016878616.1 | 5 - 6 | 1442 | 58 | |
XM_017023128.1 | XP_016878617.1 | 5 - 6 | 1247 | 58 | |
XM_017023129.1 | XP_016878618.1 | 5 - 6 | 2333 | 58 | |
XM_017023130.1 | XP_016878619.1 | 5 - 6 | 915 | 58 | |
XM_017023131.1 | XP_016878620.1 | 5 - 6 | 774 | 58 | |
XM_017023132.1 | XP_016878621.1 | 5 - 6 | 1077 | 58 | |
XM_017023133.1 | XP_016878622.1 | 5 - 6 | 1103 | 58 | |
XM_017023134.1 | XP_016878623.1 | 5 - 6 | 882 | 58 | |
GenBank mRNA | AF176701.1 | - | 5 - 6 | 772 | 58 |
AK316602.1 | - | 5 - 6 | 775 | 58 | |
BC018785.1 | - | 5 - 6 | 927 | 58 | |
BI085522.1 | - | 4 - 5 | 644 | 58 |