The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146184.1 | NP_001139656.1 | 7 - 8 | 1090 | 65 |
NM_001146185.1 | NP_001139657.1 | 7 - 8 | 776 | 65 | |
NM_001146186.1 | NP_001139658.1 | 5 - 6 | 1123 | 65 | |
NM_006210.2 | NP_006201.1 | 8 - 9 | 1166 | 65 | |
GenBank mRNA | AF208967.1 | - | 8 - 9 | 1136 | 65 |
AF208968.1 | - | 7 - 8 | 1060 | 65 | |
AF208969.1 | - | 7 - 8 | 746 | 65 | |
AF208970.1 | - | 5 - 6 | 1124 | 65 |