The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004055.1 | NP_001004055.1 | 6 - 7 | 936 | 71 |
NM_012163.2 | NP_036295.1 | 6 - 7 | 1101 | 71 | |
XM_017023133.1 | XP_016878622.1 | 6 - 7 | 1277 | 71 | |
GenBank mRNA | AK096101.1 | - | 4 - 5 | 1573 | 71 |
BC018785.1 | - | 6 - 7 | 1101 | 71 |