Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG9
Entrez Gene ID
79796
Gene Name
ALG9, alpha-1,2-mannosyltransferase
Gene Aliases
CDG1L, DIBD1, GIKANIS, LOH11CR1J
Chromosome Location
Chr.11: 111776096 - 111871581 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077690.1 NP_001071158.1 2 - 3 370 78
NM_001077691.1 NP_001071159.1 2 - 3 647 78
NM_001077692.1 NP_001071160.1 2 - 3 647 78
NM_024740.2 NP_079016.2 2 - 3 370 78
XM_005277723.4 XP_005277780.1 2 - 3 370 78
XM_005277724.4 XP_005277781.1 2 - 3 370 78
XM_006718913.2 XP_006718976.1 2 - 3 370 78
XM_011542992.1 XP_011541294.1 2 - 3 370 78
XM_017018313.1 XP_016873802.1 2 - 3 370 78
XM_017018314.1 XP_016873803.1 2 - 3 370 78
XM_017018315.1 XP_016873804.1 2 - 3 370 78
XM_017018316.1 XP_016873805.1 2 - 3 667 78
XM_017018317.1 XP_016873806.1 2 - 3 667 78
XM_017018319.1 XP_016873808.1 2 - 3 504 78
XM_017018321.1 XP_016873810.1 2 - 3 510 78
XM_017018323.1 XP_016873812.1 2 - 3 667 78
XM_017018324.1 XP_016873813.1 2 - 3 654 78
XM_017018325.1 XP_016873814.1 2 - 3 667 78
GenBank mRNA AF395532.1 - 2 - 3 322 78
AF454937.1 - 2 - 3 281 78
AK025498.1 - 2 - 3 370 78
AK172828.1 - 2 - 3 231 78
AK225261.1 - 2 - 3 349 78
AK308025.1 - 2 - 3 336 78
AL136927.1 - 2 - 3 293 78
BC009255.2 - 2 - 3 319 78
KU178783.1 - 2 - 3 272 78
KU178784.1 - 2 - 3 272 78

View Full Product Details