Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS9
Entrez Gene ID
360200
Gene Name
transmembrane protease, serine 9
Gene Aliases
-
Chromosome Location
Chr.19: 2385254 - 2426261 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182973.1 NP_892018.1 9 - 10 1474 61
XM_011527978.2 XP_011526280.1 11 - 12 1837 61
XM_011527980.1 XP_011526282.1 3 - 4 493 61
GenBank mRNA AJ488946.1 - 9 - 10 1474 61
AJ488947.1 - 9 - 10 1474 61
AK131261.1 - 1 - 2 63 61

View Full Product Details