The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014589.2 | NP_055404.1 | 2 - 3 | 239 | 125 |
GenBank mRNA | AF189279.1 | - | 2 - 3 | 239 | 125 |
BC069116.1 | - | 2 - 3 | 239 | 125 |