The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001896.2 | NP_001887.1 | 3 - 4 | 457 | 128 |
XM_005255801.3 | XP_005255858.1 | 3 - 4 | 453 | 128 | |
GenBank mRNA | AI150700.1 | - | 3 - 4 | 457 | 128 |
BC008812.2 | - | 3 - 4 | 331 | 128 | |
M55268.1 | - | 4 - 5 | 478 | 128 |