The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001896.2 | NP_001887.1 | 5 - 6 | 570 | 85 |
XM_005255800.3 | XP_005255857.1 | 3 - 4 | 655 | 85 | |
XM_005255801.3 | XP_005255858.1 | 5 - 6 | 566 | 85 | |
GenBank mRNA | BC008812.2 | - | 5 - 6 | 444 | 85 |
M55268.1 | - | 6 - 7 | 591 | 85 |