Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEP1A
Entrez Gene ID
4224
Gene Name
meprin A subunit alpha
Gene Aliases
PPHA
Chromosome Location
Chr.6: 46793357 - 46845987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005588.2 NP_005579.2 3 - 4 187 73
XM_011514628.1 XP_011512930.1 2 - 3 244 73
XM_011514629.1 XP_011512931.1 3 - 4 187 73
GenBank mRNA AF478685.1 - 3 - 4 155 73
AK225212.1 - 3 - 4 177 73
AK290282.1 - 3 - 4 155 73
AK315246.1 - 3 - 4 159 73
AK315840.1 - 3 - 4 157 73
BC131714.1 - 3 - 4 158 73
BC143650.1 - 1 - 2 326 73
BC143651.1 - 2 - 3 237 73
DA432559.1 - 3 - 4 187 73
M82962.1 - 3 - 4 154 73

View Full Product Details