The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307924.1 | NP_001294853.1 | 10 - 11 | 1404 | 102 |
NM_004537.5 | NP_004528.1 | 10 - 11 | 1404 | 102 | |
NM_139207.3 | NP_631946.1 | 10 - 11 | 1404 | 102 | |
XM_006719425.3 | XP_006719488.1 | 8 - 9 | 867 | 102 | |
XM_011538390.2 | XP_011536692.2 | 10 - 11 | 1063 | 102 | |
XM_011538393.2 | XP_011536695.1 | 10 - 11 | 1316 | 102 | |
XM_017019338.1 | XP_016874827.1 | 10 - 11 | 1316 | 102 | |
XM_017019339.1 | XP_016874828.1 | 8 - 9 | 1127 | 102 | |
XM_017019340.1 | XP_016874829.1 | 8 - 9 | 867 | 102 | |
GenBank mRNA | AK122670.1 | - | 8 - 9 | 938 | 102 |
AK295147.1 | - | 10 - 11 | 983 | 102 | |
AK297500.1 | - | 7 - 8 | 876 | 102 | |
AK304787.1 | - | 9 - 10 | 1017 | 102 | |
AL162068.1 | - | 10 - 11 | 1041 | 102 | |
BC002387.2 | - | 10 - 11 | 1038 | 102 | |
BC015599.1 | - | 9 - 10 | 1253 | 102 | |
BT007023.1 | - | 9 - 10 | 917 | 102 | |
M86667.1 | - | 10 - 11 | 992 | 102 |