The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021916.3 | NP_068735.1 | 1 - 2 | 378 | 141 |
GenBank mRNA | BC040161.1 | - | 1 - 2 | 378 | 141 |