Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMS2CL
Entrez Gene ID
441194
Gene Name
PMS2 C-terminal like pseudogene
Gene Aliases
PMS2P13
Chromosome Location
Chr.7: 6735305 - 6751601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC041364.1 - 5 - 6 563 135
CR749616.1 - 2 - 3 147 135

View Full Product Details