The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031740.2 | NP_001026910.1 | 2 - 3 | 742 | 144 |
NM_001113482.1 | NP_001106954.1 | 2 - 3 | 742 | 144 | |
GenBank mRNA | AB188490.1 | - | 2 - 3 | 961 | 144 |
AK301399.1 | - | 2 - 3 | 336 | 144 | |
BC063587.1 | - | 2 - 3 | 199 | 144 | |
BC077730.1 | - | 2 - 3 | 742 | 144 |