The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182592.2 | NP_872398.2 | 1 - 2 | 92 | 122 |
XM_011513679.2 | XP_011511981.1 | 1 - 2 | 5319 | 122 | |
GenBank mRNA | AK096895.1 | - | 1 - 2 | 92 | 122 |
BC103995.1 | - | 1 - 2 | 92 | 122 | |
BC103996.1 | - | 1 - 2 | 92 | 122 |