The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320755.1 | NP_001307684.1 | 18 - 19 | 2506 | 175 |
NM_003170.4 | NP_003161.2 | 17 - 18 | 2404 | 175 | |
XM_017024978.1 | XP_016880467.1 | 19 - 20 | 2565 | 175 | |
XM_017024979.1 | XP_016880468.1 | 17 - 18 | 2642 | 175 | |
XM_017024980.1 | XP_016880469.1 | 16 - 17 | 2336 | 175 | |
GenBank mRNA | AL833607.1 | - | 18 - 19 | 2507 | 175 |
BC003696.1 | - | 2 - 3 | 279 | 175 | |
BC136522.1 | - | 17 - 18 | 2316 | 175 | |
BC136524.1 | - | 17 - 18 | 2316 | 175 | |
BC150268.1 | - | 17 - 18 | 2318 | 175 | |
D79984.1 | - | 17 - 18 | 2318 | 175 |