The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320755.1 | NP_001307684.1 | 4 - 5 | 547 | 112 |
NM_003170.4 | NP_003161.2 | 3 - 4 | 445 | 112 | |
XM_017024978.1 | XP_016880467.1 | 5 - 6 | 606 | 112 | |
XM_017024979.1 | XP_016880468.1 | 3 - 4 | 683 | 112 | |
XM_017024980.1 | XP_016880469.1 | 2 - 3 | 377 | 112 | |
GenBank mRNA | AL833607.1 | - | 4 - 5 | 548 | 112 |
BC136522.1 | - | 3 - 4 | 357 | 112 | |
BC136524.1 | - | 3 - 4 | 357 | 112 | |
BC150268.1 | - | 3 - 4 | 359 | 112 | |
D79984.1 | - | 3 - 4 | 359 | 112 |