The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320755.1 | NP_001307684.1 | 10 - 11 | 1396 | 157 |
NM_003170.4 | NP_003161.2 | 9 - 10 | 1294 | 157 | |
XM_017024978.1 | XP_016880467.1 | 11 - 12 | 1455 | 157 | |
XM_017024979.1 | XP_016880468.1 | 9 - 10 | 1532 | 157 | |
XM_017024980.1 | XP_016880469.1 | 8 - 9 | 1226 | 157 | |
GenBank mRNA | AL833607.1 | - | 10 - 11 | 1397 | 157 |
BC033074.1 | - | 6 - 7 | 822 | 157 | |
BC136522.1 | - | 9 - 10 | 1206 | 157 | |
BC136524.1 | - | 9 - 10 | 1206 | 157 | |
BC150268.1 | - | 9 - 10 | 1208 | 157 | |
D79984.1 | - | 9 - 10 | 1208 | 157 |