Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PODNL1
Entrez Gene ID
79883
Gene Name
podocan like 1
Gene Aliases
SLRR5B
Chromosome Location
Chr.19: 13931187 - 13953392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024825.3 NP_079101.3 1 - 2 300 69
XM_006722901.1 XP_006722964.1 1 - 2 233 69
XM_006722902.1 XP_006722965.1 1 - 2 233 69
XM_006722903.1 XP_006722966.1 1 - 2 233 69
XM_011528309.2 XP_011526611.1 1 - 2 246 69
XM_011528310.2 XP_011526612.1 1 - 2 245 69
XM_011528311.1 XP_011526613.1 1 - 2 233 69
XM_011528314.1 XP_011526616.1 1 - 2 233 69
GenBank mRNA AK290731.1 - 1 - 2 233 69
BC057786.1 - 1 - 2 300 69

View Full Product Details