Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PODNL1
Entrez Gene ID
79883
Gene Name
podocan like 1
Gene Aliases
SLRR5B
Chromosome Location
Chr.19: 13931187 - 13953392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146254.1 NP_001139726.1 4 - 5 661 87
NM_024825.3 NP_079101.3 4 - 5 685 87
XM_006722901.1 XP_006722964.1 4 - 5 618 87
XM_011528308.1 XP_011526610.1 4 - 5 652 87
GenBank mRNA AK290731.1 - 4 - 5 618 87
AK298489.1 - 4 - 5 661 87
BC057786.1 - 4 - 5 685 87

View Full Product Details