The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146254.1 | NP_001139726.1 | 5 - 6 | 764 | 84 |
NM_024825.3 | NP_079101.3 | 5 - 6 | 788 | 84 | |
XM_006722901.1 | XP_006722964.1 | 5 - 6 | 721 | 84 | |
XM_011528308.1 | XP_011526610.1 | 5 - 6 | 755 | 84 | |
XM_011528309.2 | XP_011526611.1 | 4 - 5 | 669 | 84 | |
XM_011528313.1 | XP_011526615.1 | 4 - 5 | 690 | 84 | |
GenBank mRNA | AK296101.1 | - | 3 - 4 | 484 | 84 |
AK298489.1 | - | 5 - 6 | 764 | 84 | |
BC057786.1 | - | 5 - 6 | 788 | 84 |