The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272046.1 | NP_001258975.1 | 7 - 8 | 1029 | 75 |
NM_001320804.1 | NP_001307733.1 | 7 - 8 | 971 | 75 | |
XM_011539754.2 | XP_011538056.1 | 8 - 9 | 1206 | 75 | |
XM_011539757.2 | XP_011538059.1 | 7 - 8 | 1301 | 75 | |
XM_017016177.1 | XP_016871666.1 | 7 - 8 | 781 | 75 | |
XM_017016178.1 | XP_016871667.1 | 7 - 8 | 781 | 75 | |
XM_017016179.1 | XP_016871668.1 | 7 - 8 | 734 | 75 | |
XM_017016181.1 | XP_016871670.1 | 7 - 8 | 778 | 75 | |
GenBank mRNA | AJ536328.2 | - | 7 - 8 | 953 | 75 |
AK122716.1 | - | 7 - 8 | 1024 | 75 | |
AK127756.1 | - | 7 - 8 | 860 | 75 | |
AY572972.1 | - | 7 - 8 | 791 | 75 | |
BC128588.1 | - | 6 - 7 | 793 | 75 | |
BC143380.1 | - | 7 - 8 | 731 | 75 |