Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP27
Entrez Gene ID
201176
Gene Name
Rho GTPase activating protein 27
Gene Aliases
CAMGAP1, PP905, SH3D20, SH3P20
Chromosome Location
Chr.17: 45393902 - 45432916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282290.1 NP_001269219.1 4 - 5 1348 102
NM_199282.2 NP_954976.1 4 - 5 756 102
XM_005257128.3 XP_005257185.1 4 - 5 871 102
XM_006721745.2 XP_006721808.1 7 - 8 1730 102
XM_006721747.2 XP_006721810.1 7 - 8 1730 102
XM_006721750.3 XP_006721813.1 7 - 8 1703 102
XM_011524468.1 XP_011522770.1 7 - 8 1730 102
XM_011524469.1 XP_011522771.1 7 - 8 1730 102
XM_011524470.2 XP_011522772.1 7 - 8 1703 102
XM_011524471.1 XP_011522773.1 7 - 8 1559 102
XM_011524472.2 XP_011522774.1 4 - 5 871 102
XM_011524473.2 XP_011522775.1 7 - 8 1703 102
XM_011524474.1 XP_011522776.1 4 - 5 741 102
XM_017024311.1 XP_016879800.1 5 - 6 1664 102
XM_017024312.1 XP_016879801.1 4 - 5 869 102
XM_017024313.1 XP_016879802.1 7 - 8 1703 102
GenBank mRNA AK125535.1 - 4 - 5 741 102
AK300880.1 - 4 - 5 828 102
BC101388.1 - 3 - 4 524 102
BC101389.1 - 3 - 4 524 102
BC101390.1 - 3 - 4 524 102

View Full Product Details