The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282290.1 | NP_001269219.1 | 5 - 6 | 1437 | 82 |
NM_199282.2 | NP_954976.1 | 5 - 6 | 845 | 82 | |
XM_006721745.2 | XP_006721808.1 | 8 - 9 | 1819 | 82 | |
XM_006721750.3 | XP_006721813.1 | 8 - 9 | 1792 | 82 | |
XM_011524468.1 | XP_011522770.1 | 8 - 9 | 1819 | 82 | |
XM_011524470.2 | XP_011522772.1 | 8 - 9 | 1792 | 82 | |
XM_011524471.1 | XP_011522773.1 | 8 - 9 | 1648 | 82 | |
XM_011524472.2 | XP_011522774.1 | 5 - 6 | 960 | 82 | |
XM_011524473.2 | XP_011522775.1 | 8 - 9 | 1792 | 82 | |
XM_011524474.1 | XP_011522776.1 | 5 - 6 | 830 | 82 | |
XM_017024311.1 | XP_016879800.1 | 6 - 7 | 1753 | 82 | |
XM_017024312.1 | XP_016879801.1 | 5 - 6 | 958 | 82 | |
XM_017024313.1 | XP_016879802.1 | 8 - 9 | 1792 | 82 | |
GenBank mRNA | AK125535.1 | - | 5 - 6 | 830 | 82 |
BC101388.1 | - | 4 - 5 | 613 | 82 | |
BC101389.1 | - | 4 - 5 | 613 | 82 | |
BC101390.1 | - | 4 - 5 | 613 | 82 | |
BC101391.2 | - | 3 - 4 | 532 | 82 |