Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP17L3
Entrez Gene ID
645836
Gene Name
ubiquitin specific peptidase 17-like family member 3
Gene Aliases
USP17B, USP17F
Chromosome Location
Chr.8: 7976393 - 7977985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256871.1 NP_001243800.1 1 - 1 315 119

Gene Symbol
USP17L1
Entrez Gene ID
401447
Gene Name
ubiquitin specific peptidase 17-like family member 1
Gene Aliases
USP17L1P
Chromosome Location
Chr.8: 7332387 - 7333979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256873.1 NP_001243802.1 1 - 1 315 119

Gene Symbol
USP17L2
Entrez Gene ID
377630
Gene Name
ubiquitin specific peptidase 17-like family member 2
Gene Aliases
DUB-3, DUB3, USP17
Chromosome Location
Chr.8: 12137168 - 12138760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201402.2 NP_958804.2 1 - 1 119
GenBank mRNA AY509884.1 - 1 - 1 119

Gene Symbol
USP17L4
Entrez Gene ID
645402
Gene Name
ubiquitin specific peptidase 17-like family member 4
Gene Aliases
-
Chromosome Location
Chr.8: 7337115 - 7338707 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256874.1 NP_001243803.1 1 - 1 315 119

View Full Product Details