Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA12
Entrez Gene ID
353324
Gene Name
spermatogenesis associated 12
Gene Aliases
SRG5
Chromosome Location
Chr.3: 57060132 - 57076068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181727.1 NP_859078.1 2 - 2 1949 132
XM_011533676.2 XP_011531978.1 4 - 4 2495 132
GenBank mRNA AY221117.1 - 2 - 2 1949 132
BC033337.1 - 2 - 2 1724 132

View Full Product Details