Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SSX4
- Entrez Gene ID
-
6759
- Gene Name
- SSX family member 4
- Gene Aliases
-
CT5.4
- Chromosome Location
-
Chr.X: 48383527 - 48393343 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175729.1 |
NP_783856.1 |
4 - 5 |
336 |
101 |
- Gene Symbol
- SSX4B
- Entrez Gene ID
-
548313
- Gene Name
- SSX family member 4B
- Gene Aliases
-
CT5.4
- Chromosome Location
-
Chr.X: 48402082 - 48411910 on Build GRCh38
View Full Product Details