The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017398.1 | NP_001017398.1 | 1 - 2 | 578 | 90 |
GenBank mRNA | AI142389.1 | - | 1 - 2 | 335 | 90 |
BF592910.1 | - | 1 - 2 | 90 |