The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178510.1 | NP_848605.1 | 8 - 8 | 2339 | 64 |
XM_011542736.2 | XP_011541038.1 | 9 - 9 | 4149 | 64 | |
XM_011542737.2 | XP_011541039.1 | 8 - 8 | 4119 | 64 | |
XM_011542738.2 | XP_011541040.1 | 9 - 9 | 4407 | 64 | |
XM_017017475.1 | XP_016872964.1 | 9 - 9 | 4146 | 64 | |
GenBank mRNA | AF487542.1 | - | 8 - 8 | 2245 | 64 |
AJ541797.2 | - | 8 - 8 | 2339 | 64 |