The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004514.3 | NP_004505.2 | 9 - 9 | 3122 | 145 |
GenBank mRNA | AA889932.1 | - | 1 - 1 | 145 | |
AK098140.1 | - | 1 - 1 | 1940 | 145 |