Product Details

Assay Reference Genome
Location

Chr.19:18785682 on build GRCh38
Cytoband
19p13.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1311
Gene Symbol
COMP
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
Location
Chr.19:18782773-18791305 on build GRCh38
Assay Gene Location
Overlaps Intron 14 - Exon 14
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
COMP NM_000095.2 NP_000086.2
AB086984.1 BAC53888.1
AK074508.1 BAC11031.1
AK223216.1 BAD96936.1
AK290595.1
AK296586.1
AK297798.1
AK297814.1
BC110847.1 AAI10848.1
BC125092.1
L32137.1 AAA57253.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1151217 Chr19:18627369 - 19284914 on Build GRCh38 Deletion BORCS8 KLHL26 COMP CRTC1 GDF1 SUGP2 UPF1 HAPLN4 NCAN CERS1 TMEM161A SUGP1 SLC25A42 MEF2B BORCS8-MEF2B NR2C2AP RFXANK LOC102724360 ARMC6 DDX49 COPE TM6SF2 HOMER3
nsv952422 Chr19:18606691 - 18807891 on Build GRCh38 Deletion KLHL26 COMP TMEM59L CRTC1 CRLF1
nsv833777 Chr19:18765126 - 18920267 on Build GRCh38 Loss COMP CRTC1 GDF1 UPF1 CERS1 DDX49 COPE

View Full Product Details