The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243042.1 | NP_001229971.1 | 5 - 6 | 1077 | 84 |
NM_002117.5 | NP_002108.4 | 5 - 6 | 1077 | 84 | |
GenBank mRNA | AJ010749.1 | - | 5 - 6 | 1012 | 84 |
AK130592.1 | - | 6 - 7 | 943 | 84 | |
AK301283.1 | - | 5 - 6 | 1141 | 84 | |
AK304527.1 | - | 5 - 6 | 889 | 84 | |
BC007814.2 | - | 5 - 6 | 1010 | 84 | |
BC033293.1 | - | 5 - 6 | 1019 | 84 | |
D38526.1 | - | 5 - 6 | 1012 | 84 | |
D49552.1 | - | 5 - 6 | 1012 | 84 | |
D49819.1 | - | 5 - 6 | 1012 | 84 | |
M11886.1 | - | 4 - 5 | 940 | 84 | |
U09853.1 | - | 5 - 6 | 1012 | 84 | |
X83394.1 | - | 5 - 6 | 1033 | 84 | |
X97321.1 | - | 5 - 6 | 1012 | 84 |